ENA432540249 Enamine

3-(3-chlorophenyl)-3-fluoropyrrolidine hydrochloride

  • MDL number MFCD28145639
  • Cas Number : 1803592-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-199472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199472
Availability details for
Quantity
  • Availability
  • Add to Cart