ENA432540253 Enamine

1-{[3-(thiophen-2-yl)-1,2,4-oxadiazol-5-yl]methyl}piperazine hydrochloride

  • MDL number MFCD28145655
  • Cas Number : 1803597-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-34571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34571
Availability details for
Quantity
  • Availability
  • Add to Cart