ENA432540254 Enamine

1-[(3-methyl-1,2,4-oxadiazol-5-yl)methyl]piperazine hydrochloride

  • MDL number MFCD28145656
  • Cas Number : 1803580-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34850
Availability details for
Quantity
  • Availability
  • Add to Cart