ENA432540256 Enamine

tert-butyl N-[2-(cyclopentylmethyl)-3-hydroxypropyl]carbamate

  • MDL number MFCD28145559
  • Cas Number : 1697762-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198667
Availability details for
Quantity
  • Availability
  • Add to Cart