ENA432540259 Enamine

3-(2-aminoethyl)benzonitrile hydrochloride

  • MDL number MFCD21362292
  • Cas Number : 172348-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198670
Availability details for
Quantity
  • Availability
  • Add to Cart