ENA432540260 Enamine

3-(2-aminoethyl)benzonitrile

  • MDL number MFCD19207366
  • Cas Number : 219919-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-198671

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P310,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
hcodes C,T,Xn
MSDS https://www.enaminestore.com/catalog/EN300-198671
pictograms GHS05,GHS06
hstatements H301,H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart