ENA432540261 Enamine

(2,6-dimethyloxan-4-yl)methanamine

  • MDL number MFCD24252512
  • Cas Number : 1555719-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198673
Availability details for
Quantity
  • Availability
  • Add to Cart