ENA432540265 Enamine

(1R)-1-[4-(methylsulfanyl)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD16294237
  • Cas Number : 1391401-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198695
Availability details for
Quantity
  • Availability
  • Add to Cart