ENA432540270 Enamine

N-(2,2-difluoroethyl)-1-methanesulfonylpiperidin-4-amine hydrochloride

  • MDL number MFCD28145564
  • Cas Number : 1803581-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198902

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198902
Availability details for
Quantity
  • Availability
  • Add to Cart