ENA432540274 Enamine

[1-(dimethylamino)cyclobutyl]methanol

  • MDL number MFCD17170809
  • Cas Number : 1378667-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198909
Availability details for
Quantity
  • Availability
  • Add to Cart