ENA432540275 Enamine

N-butyl-3-hydroxybenzene-1-sulfonamide

  • MDL number MFCD11594629
  • Cas Number : 1082469-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198910
Availability details for
Quantity
  • Availability
  • Add to Cart