ENA432540278 Enamine

methyl 3-(benzylamino)oxolane-3-carboxylate hydrochloride

  • MDL number MFCD28145567
  • Cas Number : 1803610-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198924
Availability details for
Quantity
  • Availability
  • Add to Cart