ENA432540284 Enamine

3-(2-chloroethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD03085747
  • Cas Number : 41078-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68.5-69.5
Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198942
Availability details for
Quantity
  • Availability
  • Add to Cart