ENA432540288 Enamine

1,2,2-trimethylpiperazine dihydrochloride

  • MDL number MFCD26959634
  • Cas Number : 932047-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198953
Availability details for
Quantity
  • Availability
  • Add to Cart