ENA432540292 Enamine

2-acetamido-4-sulfanylbutanoic acid

  • MDL number MFCD01723057
  • Cas Number : 55585-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198964
Availability details for
Quantity
  • Availability
  • Add to Cart