ENA432540307 Enamine

2-hydroxy-5-(trifluoromethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD18397613
  • Cas Number : 1261500-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198998
Availability details for
Quantity
  • Availability
  • Add to Cart