ENA432540319 Enamine

2,3,5-trichloro-6-hydroxybenzene-1-sulfonyl chloride

  • MDL number MFCD25961982
  • Cas Number : 292823-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199012

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-199012
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart