ENA432540330 Enamine

4-fluoro-2-hydroxybenzene-1-sulfonyl chloride

  • MDL number MFCD18396833
  • Cas Number : 1261674-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199023
Availability details for
Quantity
  • Availability
  • Add to Cart