ENA432540338 Enamine

1-(1-phenyl-1H-pyrazol-5-yl)ethan-1-one

  • MDL number MFCD20486397
  • Cas Number : 114998-59-3
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1-phenyl-1H-pyrazol-5-yl)ethanone
Inchi Code 1S/C11H10N2O/c1-9(14)11-7-8-12-13(11)10-5-3-2-4-6-10/h2-8H,1H3
Inchi Key VOEPLNUVMOGCDS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198009
Availability details for
Quantity
  • Availability
  • Add to Cart