ENA432540340 Enamine

2-(chloromethyl)-1,3-difluoro-5-nitrobenzene

  • MDL number MFCD15528808
  • Cas Number : 1343441-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198019
Availability details for
Quantity
  • Availability
  • Add to Cart