ENA432540341 Enamine

methyl 3-amino-2-chloro-6-methylpyridine-4-carboxylate

  • MDL number MFCD22058645
  • Cas Number : 1369085-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198256
Availability details for
Quantity
  • Availability
  • Add to Cart