ENA432540349 Enamine

2,5-dichloro-4-hydroxybenzene-1-sulfonyl chloride

  • MDL number MFCD24329380
  • Cas Number : 71292-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-199042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199042
Availability details for
Quantity
  • Availability
  • Add to Cart