ENA432540355 Enamine

[(2-bromo-4-methylphenyl)methyl][(thiophen-2-yl)methyl]amine

  • MDL number MFCD24229505
  • Cas Number : 1546851-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199084
Availability details for
Quantity
  • Availability
  • Add to Cart