ENA432540356 Enamine

N-[(2-fluorophenyl)methyl]-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD04206902
  • Cas Number : 606136-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199085
Availability details for
Quantity
  • Availability
  • Add to Cart