ENA432540357 Enamine

5-bromo-N-[2-(1H-imidazol-1-yl)ethyl]pyridin-2-amine

  • MDL number MFCD14624951
  • Cas Number : 1248508-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199086
Availability details for
Quantity
  • Availability
  • Add to Cart