ENA432540358 Enamine

5-bromo-N-[2-(dimethylamino)ethyl]pyridin-2-amine

  • MDL number MFCD11125892
  • Cas Number : 112357-06-9
  • Molecular Weight : 244.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N~1~-(5-bromo-2-pyridinyl)-N~2~,N~2~-dimethyl-1,2-ethanediamine
Inchi Code 1S/C9H14BrN3/c1-13(2)6-5-11-9-4-3-8(10)7-12-9/h3-4,7H,5-6H2,1-2H3,(H,11,12)
Inchi Key YZNKVSIFLBFINU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199087
Availability details for
Quantity
  • Availability
  • Add to Cart