ENA432540367 Enamine

5-bromo-4-cyclopropyl-2-methylpyrimidine

  • MDL number MFCD22032112
  • Cas Number : 1368136-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198358
Availability details for
Quantity
  • Availability
  • Add to Cart