ENA432540368 Enamine

3-chloro-5-[(4-methylphenyl)methanesulfonyl]-1,2,4-thiadiazole

  • MDL number MFCD02946292
  • Cas Number : 494763-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198365
Availability details for
Quantity
  • Availability
  • Add to Cart