ENA432540369 Enamine

1-(bromomethyl)-1-methylcyclobutane

  • MDL number MFCD20627042
  • Cas Number : 98775-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-199106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199106
Availability details for
Quantity
  • Availability
  • Add to Cart