ENA432540370 Enamine

1-[5-(trifluoromethyl)pyridin-2-yl]ethan-1-ol

  • MDL number MFCD26402861
  • Cas Number : 1345973-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199110
Availability details for
Quantity
  • Availability
  • Add to Cart