ENA432540372 Enamine

(2R)-2-amino-2-(4-ethoxyphenyl)ethan-1-ol hydrochloride

  • MDL number MFCD12758152
  • Cas Number : 1391501-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-198439
Availability details for
Quantity
  • Availability
  • Add to Cart