ENA432540377 Enamine

4-amino-6-hydroxybenzene-1,3-dicarboxylic acid

  • MDL number MFCD20638466
  • Cas Number : 15540-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199121
Availability details for
Quantity
  • Availability
  • Add to Cart