ENA432540379 Enamine

2-amino-4-methoxybutan-1-ol

  • MDL number MFCD19216184
  • Cas Number : 1314909-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199124
Availability details for
Quantity
  • Availability
  • Add to Cart