ENA432540383 Enamine

8-propoxy-N-propylquinolin-4-amine

  • MDL number MFCD17312575
  • Cas Number : 1275675-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199132
Availability details for
Quantity
  • Availability
  • Add to Cart