ENA432540386 Enamine

1-(piperazin-1-yl)but-3-en-1-one; trifluoroacetic acid

  • MDL number MFCD28145606
  • Cas Number : 1803601-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199135
Availability details for
Quantity
  • Availability
  • Add to Cart