ENA432540390 Enamine

[3-(benzyloxy)propyl](methyl)amine

  • MDL number MFCD16662989
  • Cas Number : 188878-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199141
Availability details for
Quantity
  • Availability
  • Add to Cart