ENA432540392 Enamine

1-(hydroxymethyl)cyclohexan-1-ol

  • MDL number MFCD00956115
  • Cas Number : 15753-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-76
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199144
Availability details for
Quantity
  • Availability
  • Add to Cart