ENA432540394 Enamine

N-cyclopropyl-2-hydroxy-N-methylacetamide

  • MDL number MFCD14678337
  • Cas Number : 1247116-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199146
Availability details for
Quantity
  • Availability
  • Add to Cart