ENA432540400 Enamine

1-(2,3-dihydro-1,4-benzodioxin-2-yl)-2-phenylethan-1-amine

  • MDL number MFCD20795782
  • Cas Number : 1485392-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198475
Availability details for
Quantity
  • Availability
  • Add to Cart