ENA432540402 Enamine

1-(3-methoxyphenyl)ethane-1-sulfonamide

  • MDL number MFCD16672023
  • Cas Number : 1250487-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199249
Availability details for
Quantity
  • Availability
  • Add to Cart