ENA432540404 Enamine

1-(4-chlorophenyl)-4-methyl-1H-pyrazol-3-amine hydrochloride

  • MDL number MFCD28145556
  • Cas Number : 1803610-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198501
Availability details for
Quantity
  • Availability
  • Add to Cart