ENA432540405 Enamine

2-sulfamoylbenzamide

  • MDL number MFCD01365825
  • Cas Number : 36547-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198511
Availability details for
Quantity
  • Availability
  • Add to Cart