ENA432540407 Enamine

methyl 3-amino-2-(propan-2-yloxy)benzoate hydrochloride

  • MDL number MFCD28145557
  • Cas Number : 1803581-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198538
Availability details for
Quantity
  • Availability
  • Add to Cart