ENA432540412 Enamine

2-benzylimidazo[1,2-a]pyridine

  • MDL number MFCD24199319
  • Cas Number : 889813-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199282
Availability details for
Quantity
  • Availability
  • Add to Cart