ENA432540413 Enamine

octahydropyrrolo[1,2-a]piperazin-3-one

  • MDL number MFCD17926435
  • Cas Number : 16620-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199283
Availability details for
Quantity
  • Availability
  • Add to Cart