ENA432540414 Enamine

2,2-dimethylpropane-1-thiol

  • MDL number MFCD00603028
  • Cas Number : 1679-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-198544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198544
Availability details for
Quantity
  • Availability
  • Add to Cart