ENA432540417 Enamine

methyl 2-amino-3-nitropyridine-4-carboxylate

  • MDL number MFCD28145558
  • Cas Number : 1803592-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198626
Availability details for
Quantity
  • Availability
  • Add to Cart