ENA432540418 Enamine

ethyl 2-(2-oxoazepan-3-yl)acetate

  • MDL number MFCD24570533
  • Cas Number : 831-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198632
Availability details for
Quantity
  • Availability
  • Add to Cart