ENA432540422 Enamine

2-(1-benzylpyrrolidin-3-yl)pyridine

  • MDL number MFCD11042353
  • Cas Number : 145105-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198637
Availability details for
Quantity
  • Availability
  • Add to Cart