ENA432540426 Enamine

methyl 5-(bromomethyl)-1-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD21362488
  • Cas Number : 878744-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199308
Availability details for
Quantity
  • Availability
  • Add to Cart